missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse GPT His Protein

Product Code. p-7111133 Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
This item is not returnable. View return policy

Product Code. 18292521

Brand: Novus Biologicals™ NBP25954920UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified and high bioactivity. Generating reliable and reproducible results. Applications: Functional, SDS-Page, Bioactivity

A bioactive recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-496 of Mouse GPT The Recombinant Mouse GPT Protein is derived from E. coli. The Recombinant Mouse GPT Protein has been validated for the following applications: Functional, SDS-Page, Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1mg/mL
For Use With (Application) SDS-PAGE
Formulation Liquid. PBS (pH7.4) containing 1mM DTT, 10% glycerol
Gene ID (Entrez) 2875
Molecular Weight (g/mol) 57.5 kDa
Name Recombinant Mouse GPT Protein
Purification Method >90%, by SDS-PAGE
Quantity 20 μg
Source E.coli
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Regulatory Status RUO
Gene Alias AAT1alanine aminotransferase 1, ALT1EC 2.6.1.2, Glutamate pyruvate transaminase 1, glutamic-alanine transaminase 1, Glutamic--alanine transaminase 1, glutamic-pyruvate transaminase (alanine aminotransferase), Glutamic--pyruvic transaminase 1, GPT1GPT 1
Gene Symbol GPT
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Mouse
Recombinant Recombinant
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.