missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GOLM1 / GP73 Protein
A DNA sequence encoding the Mus musculus (Mouse) GOLM1 / GP73, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6106-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001030294.1 | |
GOLM1 / GP73 Protein | |
Research Use Only | |
GOLM1 | |
Recombinant Protein | |
E. coli | |
SSRSVELQTR IVELEGRVRR AAAERGAVEL KKNEFQGELQ KQREQLDRIQ SSHSFQLENV NKLHQDEKAV LVNNITTGEK LIRDLQDQLK ALQRSYSSLQ QDIFQFQKNQ TSLEKKFSYD LNQCISQMTE VKEQCDERIE EVIRKRNEAP GSRDLAETNN QHQQALKPQP KLQEEVPSEE QMPQEKGDVP RNKSQIPAPN SESLGLKPQV QNEETNEIQA VGEEHQQASI QGMS-QAVAD GTRVGAEKLD QHTQLPAGLL ARPEEDSQYP EREQLVIRDR QEQQRASEEG GGQKNPGDEY DMDENEAESE REKQAALAGN DRNINVLNAD AQKRGIINVP VGSERQSHIL NQVGIHIPQQ A | |
Greater than 90% as determined by SDS-PAGE. |
105348 | |
10 μg | |
Not determined. | |
Full Length of Mature Protein | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |