missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse GOLM1 / GP73 Protein

A DNA sequence encoding the Mus musculus (Mouse) GOLM1 / GP73, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6106-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030840

  • £560.00 / 100µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001030294.1
GOLM1 / GP73 Protein
Research Use Only
GOLM1
Recombinant Protein
E. coli
SSRSVELQTR IVELEGRVRR AAAERGAVEL KKNEFQGELQ KQREQLDRIQ SSHSFQLENV NKLHQDEKAV LVNNITTGEK LIRDLQDQLK ALQRSYSSLQ QDIFQFQKNQ TSLEKKFSYD LNQCISQMTE VKEQCDERIE EVIRKRNEAP GSRDLAETNN QHQQALKPQP KLQEEVPSEE QMPQEKGDVP RNKSQIPAPN SESLGLKPQV QNEETNEIQA VGEEHQQASI QGMS-QAVAD GTRVGAEKLD QHTQLPAGLL ARPEEDSQYP EREQLVIRDR QEQQRASEEG GGQKNPGDEY DMDENEAESE REKQAALAGN DRNINVLNAD AQKRGIINVP VGSERQSHIL NQVGIHIPQQ A
Greater than 90% as determined by SDS-PAGE.
105348
100 μg
Not determined.
Full Length of Mature Protein
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers