missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Glyoxylate reductase / hydroxypyruvate reductase Protein
A DNA sequence encoding the Mus musculus (Mouse) Glyoxylate reductase/hydroxypyruvate reductase, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_525028.1 |
---|---|
Gene ID (Entrez) | 76238 |
Name | Glyoxylate reductase / hydroxypyruvate reductase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16050940
|
enQuireBio™
QP6119-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16070940
|
enQuireBio™
QP6119-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16060940
|
enQuireBio™
QP6119-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16080940
|
enQuireBio™
QP6119-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16090940
|
enQuireBio™
QP6119-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16000950
|
enQuireBio™
QP6119-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_525028.1 | |
Glyoxylate reductase / hydroxypyruvate reductase Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
76238 | |
Research Use Only | |
Glxr | |
Recombinant Protein | |
E. coli | |
MKPARLMKVF VTGPLPAEGR AALAQAADCE VEQWNSDDPI PRKDLEQGVV GAHGLLCRLS DRVDKKLLDA AGANLRVIST LSVGVDHLAL DEIKKRGIRV GYTPGVLTDA TAELAVSLLL TTCRRLPEAI EEVKNGGWSS WSPLWMCGYG LSQSTVGIVG LGRIGQAIAR RLKPFGVQRF LYTGRQPRPQ EAAEFQAEFV PIAQLAAESD FIVVSCSLTP DTMGLCSKDF FQKMKNTAIF INISRGDVVN QEDLYQALAS GQIAAAGLDV TTPEPLPPSH PLLTLKNCVI LPHIGSATYK TRNTMSLLAA NNLLAGLRGE AMPSELKL | |
Greater than 90% as determined by SDS-PAGE. |