missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse GLO1 Protein
A cDNA sequence encoding the GLO1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10645-10ug
Additional Details : Weight : 0.01000kg
Specifications
109801 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 210 units/mg, and is defined as the amount of enzyme that will form 1.0 micromol of S-lactoylglutathione from methylglyoxal and reduced glutathione per minute at pH 6.5 at 25C. | |
Mouse | |
His | |
GLO1 protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
GLO1 Protein | |
Research Use Only | |
Glo1 | |
Recombinant Protein | |
Baculovirus | |
MAEPQPASSG LTDETAFSCC SDPDPSTKDF LLQQTMLRIK DPKKSLDFYT RVLGLTLLQK LDFPAMKFSL YFLAYEDKND IPKDKSEKTA WTFSRKATLE LTHNWGTEDD ETQSYHNGNS DPRGFGHIGI AVPDVYSACK RFEELGVKFV KKPDDGKMKG LAFIQDPDGY WIEILNPNKI ATIILEHHHH HH | |
Greater than 95.0% as determined by SDS-PAGE. |