missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse GLO1 Protein

A cDNA sequence encoding the GLO1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10645-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934508

  • £1320.00 / 100µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

109801
100 μg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity is > 210 units/mg, and is defined as the amount of enzyme that will form 1.0 micromol of S-lactoylglutathione from methylglyoxal and reduced glutathione per minute at pH 6.5 at 25C.
Mouse
His
GLO1 protein solution (0.5 mg/ml) containing Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
GLO1 Protein
Research Use Only
Glo1
Recombinant Protein
Baculovirus
MAEPQPASSG LTDETAFSCC SDPDPSTKDF LLQQTMLRIK DPKKSLDFYT RVLGLTLLQK LDFPAMKFSL YFLAYEDKND IPKDKSEKTA WTFSRKATLE LTHNWGTEDD ETQSYHNGNS DPRGFGHIGI AVPDVYSACK RFEELGVKFV KKPDDGKMKG LAFIQDPDGY WIEILNPNKI ATIILEHHHH HH
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers