missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse GFR alpha-like Fc Chimera Protein
Description
The Recombinant Mouse GFR alpha-like Fc Chimera Protein is derived from CHO. The Recombinant Mouse GFR alpha-like Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Accession Number | Q6SJE0-1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. |
| Molecular Weight (g/mol) | 64 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line,CHO-derived mouse GFR alpha-like protein, (N-terminus) Mouse GFR alpha-like (Gln20-Gly349) Accession &Num; Q6SJE0-1 IEGRMDP Mouse IgG2a (Glu98-Lys330) (C-terminus) |
| Storage Requirements | 12 months from date of receipt, ≤ -20° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, ≤ -20° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?