missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse GDF-8 Propeptide Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£359.60
Specifications
Gene ID (Entrez) | 17700 |
---|---|
Molecular Weight (g/mol) | 28.7 kDa |
Name | GDF-8/Myostatin Propeptide |
Quantity | 25μg |
Source | Mouse myeloma cell line,NS0-derived mouse GDF-8/Myostatin protein Asn25-Ser265,with a C-terminal 10-His tag |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18177440
|
R&D Systems™
1539-PG-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£359.60
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18188640
|
R&D Systems™
1539-PG-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£359.60
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Mouse GDF-8 Propeptide Protein is derived from NS0. The Recombinant Mouse GDF-8 Propeptide Protein has been validated for the following applications: Bioactivity.Specifications
17700 | |
GDF-8/Myostatin Propeptide | |
Mouse myeloma cell line,NS0-derived mouse GDF-8/Myostatin protein Asn25-Ser265,with a C-terminal 10-His tag | |
GDF8, GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, MSLHP, MSTN, Myostatin | |
Recombinant |
28.7 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
90%, by SDS-PAGE under reducing conditions and visualized by silver stain. |