missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse GDF-8 Propeptide Fc Chimera Protein

Product Code. 18349207
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in MES, NaCl and CHAPS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in MES, NaCl and CHAPS.
Unit Size:
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation unitSize
18349207 Lyophilized from a 0.2μm filtered solution in MES, NaCl and CHAPS. 25µg
18348377 Lyophilized from a 0.2μm filtered solution in MES, NaCl and CHAPS with BSA as a carrier protein. 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18349207 Supplier R&D Systems™ Supplier No. 6986PG025/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse GDF-8 Propeptide Fc Chimera Protein is derived from NS0. The Recombinant Mouse GDF-8 Propeptide Fc Chimera Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Spezifikation

Formulation Lyophilized from a 0.2μm filtered solution in MES, NaCl and CHAPS.
Gene ID (Entrez) 17700
Molecular Weight (g/mol) 54.9 kDa (monomer)
Name GDF-8/Myostatin Propeptide Fc Chimera
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived mouse GDF-8/Myostatin protein, (N-terminus) Mouse GDF-8 Propeptide (Asn25-Arg267) with a substitution Ser265Arg Accession &Num; NP_034964 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Gene Alias GDF8, GDF-8, GDF8growth differentiation factor 8, growth/differentiation factor 8, MSLHP, MSTN, Myostatin
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.