missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse GDF-7 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£327.21
Specifications
Gene ID (Entrez) | 238057 |
---|---|
Name | GDF-7/BMP-12 |
Quantity | 10μg |
Source | E. coli-derived mouse GDF-7/BMP-12 protein Thr308-Arg453 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Product Code | Brand | Formulation | Molecular Weight (g/mol) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Molecular Weight (g/mol) | Price | Quantity & Availability | ||||
18118540
|
R&D Systems™
779-G7-010/CF |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA. | 15 kDa (monomer) |
£327.21
10µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
18126550
|
R&D Systems™
779-G7-010 |
Lyophilized from a 0.2μm filtered solution in Acetonitrile and TFA with BSA as a carrier protein. | 15 kDa |
£327.21
10µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Descripción
The Recombinant Mouse GDF-7 Protein is derived from E. coli. The Recombinant Mouse GDF-7 Protein has been validated for the following applications: Bioactivity.Especificaciones
238057 | |
10μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
GDF-7/BMP-12 | |
E. coli-derived mouse GDF-7/BMP-12 protein Thr308-Arg453 | |
Disulfide-linked homodimer | |
Recombinant |