missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse GDF-15 (CHO-expressed) Protein, CF

Product Code. 18397717
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18397717 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18397717 Supplier R&D Systems™ Supplier No. 10596GD025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Carrier-Free Recombinant Protein

Measured by its ability to activate SRE-SEAP reporter in HEK293 human embryonic kidney cells transfected with human c-Ret and human GFRAL. The ED50 for this effect is 0.4-4 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Z0J7.2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in HCl.
Gene ID (Entrez) 9518
Molecular Weight (g/mol) M.W. (Observed): 11-13 kDa, under reducing conditions,, M.W. (Theoretical): 13 kDa
Quantity 25 μg
Source Chinese Hamster Ovary cell line, CHO-derived mouse GDF-15 protein Ser189-Ala303
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias GDF15, growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDF, PDFGDF-15, PLAB, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGF-beta, PTGFBPTGF-beta
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in 4 mM HCl.
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein GDF-15
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.