missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Gas6 (Full Length) Protein
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
Description
The Recombinant Mouse Gas6 (Full Length) Protein is derived from NS0. The Recombinant Mouse Gas6 (Full Length) Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
| Accession Number | CAA42507 |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS |
| Gene ID (Entrez) | 14456 |
| Molecular Weight (g/mol) | 71kDa |
| Name | Gas6 |
| Preparation Method | Reconstitute at 250μg/mL in sterile PBS. |
| Quality Control Testing | SDS-PAGE: 78-94kDa, reducing conditions |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Gas6 protein Ala46-Pro673,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction