missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals Recombinant Mouse gamma-Synuclein Active, Monomer Protein
Specifications
Specifications
| For Use With (Application) | In vitro Assay, In vivo Assay, SDS-PAGE, Western Blot |
| Formulation | PBS pH 7.4 |
| Gene ID (Entrez) | 6623 |
| Quantity | 5 x 100 μg Vials |
| Storage Requirements | Store at -80C. Avoid freeze-thaw cycles. |
| Gene Alias | BCSG1PERSYN, Breast cancer-specific gene 1 protein, gamma-synuclein, persyn, SRPRSN, synoretin, synuclein, gamma (breast cancer-specific protein 1) |
| Gene Symbol | SNCG |
| Research Category | Alzheimers Research, Cancer, Cellular Markers, Core ESC Like Genes, Mitotic Regulators, Neuroscience, Stem Cell Markers |
| Purity or Quality Grade | >95%, by SDS-PAGE |
| Protein | gamma-Synuclein |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?