missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Galectin-4 Protein
Description
The Recombinant Mouse Galectin-4 Protein is derived from E. coli. The Recombinant Mouse Galectin-4 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS, EDTA, DTT and Trehalose. |
| Gene ID (Entrez) | 16855 |
| Molecular Weight (g/mol) | 36.2 kDa |
| Name | Galectin-4 |
| Quantity | 50 μg |
| Source | E. coli-derived mouse Galectin-4 protein Met1-Ile326 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?