missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Galectin-3 Protein

Product Code. 18717448
Click to view available options
Quantity:
1 mg
50 μg
Formulation:
Lyophilized from a 0.2 μm filtered solution in PBS, EDTA and DTT with BSA as a carrier protein.
Lyophilized from a 0.2 μm filtered solution in PBS, EDTA and DTT.
Unit Size:
1mg
50µg
This item is not returnable. View return policy

Product Code. 18717448

Brand: R&D Systems™ 1197GA01M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A New and Improved rm Galectin-3 is Now Available! It is expressed in mammalian cells with a lower endotoxin spec.

Measured by its ability to agglutinate human red blood cells. Hadari, Y.R. et al. (2000) J. Cell Sci. 113:2385. The ED50 for this effect is 5-20 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_034835
Characterization Bioactivity
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS, EDTA and DTT with BSA as a carrier protein.
Gene ID (Entrez) 3958
Molecular Weight (g/mol) M.W. (theoretical): 27.3 kDa
Quantity 1 mg
Source E. coli-derived mouse Galectin-3 protein Ala2-Ile264
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 3 months, 2°C to 8°C under sterile conditions after reconstitution.
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Alias AGE-R3, Carbohydrate-binding protein 35, CBP 35,35kDa lectin, CBP35, Gal-3, GAL3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, Galectin3, GALIG, IgE-binding protein, L29, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, LGALS3, Mac-2, Mac-2 antigen, MAC2GAL3
Gene Symbol Lgals3
Product Type Recombinant Protein
Conjugate Unconjugated
Species Mouse
Reconstitution Reconstitute at 250 μg/mL in sterile PBS containing (at least 0.1% human or bovine serum albumin.)
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Galectin-3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.