missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Galectin-1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£327.21
Specifications
Gene ID (Entrez) | 16852 |
---|---|
Molecular Weight (g/mol) | 15 kDa |
Name | Galectin-1 |
Quantity | 50μg |
Source | E. coli-derived mouse Galectin-1 protein Ala2-Glu135 |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18167820
|
R&D Systems™
1245-GA-050/CF |
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT. |
£327.21
50µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18177950
|
R&D Systems™
1245-GA-050 |
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and DTT with BSA as a carrier protein. |
£327.21
50µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Mouse Galectin-1 Protein is derived from E. coli. The Recombinant Mouse Galectin-1 Protein has been validated for the following applications: Bioactivity.Specifications
16852 | |
Galectin-1 | |
E. coli-derived mouse Galectin-1 protein Ala2-Glu135 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
15 kDa | |
50μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |