missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
G-CSF protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferation assay using murine NFS60 cells is less than 0.05 ng/mL, corresponding to a specific activity of > 2.0 x 107 IU/mg.
Specifications
Specifications
| Accession Number | P09920, P09920, P09920 |
| Concentration | LYOPH |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in 10 mM Sodium Citrate, pH 4.0, 150 mM NaCl, 0.01 % Tween-20. |
| Gene ID (Entrez) | 1440 |
| Molecular Weight (g/mol) | 18.9 kDa |
| Quantity | 1 mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/ug of G-CSF as determined by LAL method. |
| Gene Alias | C17orf33, chromosome 17 open reading frame 33, colony stimulating factor 3 (granulocyte), CSF3OS, filgrastim, G-CSF, GCSFlenograstim, granulocyte colony-stimulating factor, Lenograstim, MGC45931, pluripoietin |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?