missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Furin Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
Measured by its ability to cleave the fluorogenic peptide substrate pERTKR-AMC (Catalog # ES013 ). The specific activity is >150 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | P23188 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris, CaCl2, NaCl, Brij-35 and Glycerol. |
| Gene ID (Entrez) | 5045 |
| Molecular Weight (g/mol) | M.W. (Observed): 60-80 kDa, reducing conditions; M.W. (theoretical): 77 kDa |
| Name | Furin |
| Quantity | 10 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived mouse Furin protein Gln25-Glu714, with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction