missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse FTO His Protein

Product Code. 18326416 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326416 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326416 Supplier Novus Biologicals™ Supplier No. NBP318144

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8BGW1
Concentration 0.5 mg/ml
For Use With (Application) SDS-PAGE
Formulation Liquid. 0.2um-filtered solution in 55mM TRIS-Cl, pH 8.2, 150mM NaCl
Gene ID (Entrez) 79068
Molecular Weight (g/mol) M.W. Theoretical: 55 kDa
Quantity 10 μg
Storage Requirements Store at 4C short term. Aliquot and Store at -20°C long term. Avoid freeze-thaw cycles.
Endotoxin Concentration <1 EU/μg purified protein (LAL test; Lonza).
Gene Alias alpha-ketoglutarate-dependent dioxygenase FTO, EC 1.14.11.-, fat mass and obesity associated, Fat mass and obesity-associated protein, KIAA1752protein fto, MGC5149
Gene Symbol FTO
Research Category Lipid and Metabolism, Neuroscience
Purity or Quality Grade >90% SDS-PAGE
Protein FTO
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.