missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Fgf8 Protein

Product Code. 15926219
Click to view available options
Quantity:
1 mg
100 μg
25 μg
5 μg
500 μg
Unit Size:
100µg
1mg
25µg
500µg
5µg
This item is not returnable. View return policy

Product Code. 15926219

Brand: enQuireBio™ QP547825ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_001159834.1
Gene ID (Entrez) 14179
Name Fgf8 Protein
Quantity 25 μg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol FGF8
Biological Activity 3T3 cell proliferation significantly induced by less than 150 ng/ml of Mouse Fgf8 (typical ED50 is ? 20 ng/ml).
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag Untagged
Sequence MQVTVQSSPN . FTQHVREQS L. VTDQLSRR LI. RTYQLYS RTS. GKHVQV LANK. RINAM AEDGD. PFAK LIVETD. TFG SRVRVRGAET GLYICMN. KK GKLIAKSN. G KGKDCVFTE.  IVLENNYTAL . QNAKYEGWY M. AFTRKGRP RK. GSKTRQH QRE. VHFMKR LPRG. HHTTE QSLRFEFLNY PPFTR. SLRG SQRTWA. PEP R
Buffer 10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.