missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Fgf8 Protein
Recombinant Protein
Brand: enQuireBio™ QP5478-100ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001159834.1 | |
Fgf8 Protein | |
Research Use Only | |
FGF8 | |
Recombinant Protein | |
E. coli | |
MQVTVQSSPN . FTQHVREQS L. VTDQLSRR LI. RTYQLYS RTS. GKHVQV LANK. RINAM AEDGD. PFAK LIVETD. TFG SRVRVRGAET GLYICMN. KK GKLIAKSN. G KGKDCVFTE. IVLENNYTAL . QNAKYEGWY M. AFTRKGRP RK. GSKTRQH QRE. VHFMKR LPRG. HHTTE QSLRFEFLNY PPFTR. SLRG SQRTWA. PEP R | |
Greater than 95% as determined by SDS-PAGE. |
14179 | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
3T3 cell proliferation significantly induced by less than 150 ng/ml of Mouse Fgf8 (typical ED50 is ? 20 ng/ml). | |
Mouse | |
Untagged | |
10 mM sodium phosphate, 50 mM sodium chloride, pH 7.5 |