missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse FGF basic/FGF2/bFGF Protein

Product Code. 18384837
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18384837 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18384837 Supplier R&D Systems™ Supplier No. 3139FB025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse FGF basic/FGF2 Protein is derived from E. coli. The Recombinant Mouse FGF basic/FGF2 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 EDTA and DTT with BSA as a carrier protein.
Gene ID (Entrez) 14173
Molecular Weight (g/mol) 16.2 kDa
Name FGF basic
Quantity 25 μg
Source E. coli-derived mouse FGF basic/FGF2/bFGF protein Ala11-Ser154
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGF2, FGF-2, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2, Prostatropin
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.