missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse FGF basic/FGF2/bFGF Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
10 μg
100 μg
1mg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
Description
FGF basic/FGF2/bFGF protein is fully biologically active when compared to standard. The ED50 as determined by a cell proliferationassay using murine balb/c 3T3 cells is less than 1.0 ng/mL, corresponding to a specific activity of >1.0 x 10^6 IU/mg
Specifications
Specifications
| Accession Number | P15655, P15655, P15655 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 2247 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 16.5 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of FGF basic/FGF2/bFGF as determined by LAL method. |
| Gene Alias | Basic fibroblast growth factor, basic fibroblast growth factor bFGF, bFGF, FGFBprostatropin, fibroblast growth factor 2 (basic), HBGF-2, heparin-binding growth factor 2 |
| Gene Symbol | FGF2 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction