missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse FGF-21 Protein

Product Code. 18498891 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18498891 1 mg 1mg
18460372 500 μg 500µg
18401732 5 μg 5µg
18490242 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18498891 Supplier Novus Biologicals™ Supplier No. NBP2350311mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

FGF-21 protein is fully biologically active when compared to standard. The ED50 as determined by thymidine uptake assay using FGF-receptors transfected BaF3 cells is less than 0.5 μg/mL, corresponding to a specific activity of >2.0 x 10^3 IU/mg in the presence of 5 μg/mL of rMuKlotho-beta and 10 μg/mL of heparin.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9JJN1, Q9JJN1, Q9JJN1
Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution in 3 x PBS, pH 7.4.
Gene ID (Entrez) 26291
Molecular Weight (g/mol) 19.9 kDa
Quantity 1 mg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of FGF-21 as determined by LAL method.
Gene Alias FGF-21, fibroblast growth factor 21
Gene Symbol FGF21
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C.
Research Category Angiogenesis, Biologically Active Proteins, Cancer
Purity or Quality Grade >97% pure by SDS-PAGE and HPLC
Protein FGF-21
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.