missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Fc gamma RIIB/CD32b Protein
Description
The Recombinant Mouse Fc gamma RIIB/CD32b Protein is derived from NS0. The Recombinant Mouse Fc gamma RIIB/CD32b Protein has been validated for the following applications: Binding Activity.
Specifications
Specifications
| Accession Number | P08101 |
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 14130 |
| Molecular Weight (g/mol) | 22 kDa |
| Name | Fc gamma RIIB/CD32b |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Fc gamma RIIB/CD32b protein Thr30-Arg207,with a C-terminal 10-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Monomer |
| Conjugate | Unconjugated |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?