missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Fas/TNFRSF6/CD95 Fc Chimera Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
50µg
Description
The Recombinant Mouse Fas/TNFRSF6/CD95 Fc Chimera Protein is derived from Sf 21 (baculovirus). The Recombinant Mouse Fas/TNFRSF6/CD95 Fc Chimera Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
| Gene ID (Entrez) | 14102 |
| Molecular Weight (g/mol) | 46 kDa (monomer) |
| Name | Fas/TNFRSF6/CD95 Fc Chimera |
| Quantity | 50 μg |
| Source | Spodoptera frugiperda, Sf 21 (baculovirus)-derived mouse Fas/TNFRSF6/CD95 protein, (N-terminus) Mouse FAS (Gly14 - Arg169) Accession # P25446 DIEGRMD Human IgG1 (Pro100 - Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Structural Form | Disulfide-linked homodimer |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction