missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse Fas Ligand/TNFSF6 Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Molecular Weight (g/mol):
18 kDa
18 kDa (monomer)
Unit Size:
50µg
Description
The Recombinant Mouse Fas Ligand/TNFSF6 Protein is derived from NS0. The Recombinant Mouse Fas Ligand/TNFSF6 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 14103 |
| Molecular Weight (g/mol) | 18 kDa |
| Name | Fas Ligand/TNFSF6 |
| Quantity | 50 μg |
| Source | Mouse myeloma cell line,NS0-derived mouse Fas Ligand/TNFSF6 protein Pro132-Leu279,with an N-terminal Met and 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction