missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
liver FABP Protein is fully biologically active when compared to standard. The binding affinity of rMuFABP1 for the synthetic ligand cis-parinaric acid has been measured by fluorescence titration. Half maximal fluorescence of 2.5 uM rMuFABP1 is achieved with approximately 5 uM cis-paranaric acid.
Specifications
Specifications
| Accession Number | P12710 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4, 2 % trehalose. |
| Gene ID (Entrez) | 2168 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 14.2 kDa |
| Quantity | 500 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of FABP1/L-FABP as determined by LAL method. |
| Gene Alias | FABP1, FABPL, fatty acid binding protein 1, liver, fatty acid-binding protein, liver, L-FABPFatty acid-binding protein 1, Liver-type fatty acid-binding protein |
| Gene Symbol | FABP1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?