missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse F-Spondin/SPON1 Protein
Click to view available options
:
50ug; Carrier Free
Description
The Recombinant Mouse F-Spondin/SPON1 Protein is derived from NS0. The Recombinant Mouse F-Spondin/SPON1 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 233744 |
| Molecular Weight (g/mol) | 88.2 kDa |
| Name | F-Spondin/SPON1 |
| Quantity | 50μg |
| Source | Mouse myeloma cell line,NS0-derived mouse F-Spondin/SPON1 protein Leu27-Cys807 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Purity or Quality Grade | 90%, by SDS-PAGE with silver staining. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction