missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse F-box / WD repeat-containing protein 10 Protein

Product Code. 16000331
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 16000331

Brand: enQuireBio™ QP7709ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Mus musculus (Mouse) F-box/WD repeat-containing protein 10, was expressed in the hosts and tags indicated.

Specifications

Name F-box / WD repeat-containing protein 10 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol Fbxw10
Protein Length Partial (see sequence information for more details).
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag His-SUMO
Sequence VKWQYSSDKN KVKKSKDKEE EREETSLGDE HSRSTIQGHS LKDSVSSKQE FSKSRVHLKQ TKNLSSDDME TPVGEVSHPL QKLWKVPMTP DRFLLTISAL QQAHNSEEFA YPHRPRPQVI DAWGPSIPYP RKVLSLKGKS VQHAVDQLRS SNLPTGVRQT NIPLEIQKLQ PNLKKSLHSP RVQATVPQPS LIRPKVSDSL RGDEHLTSSI DGTMRRAGPL TSMQVIKPNR MLAPRGGTAT LSPKKERPRF YTTLDPLRMN TGFMLMTVKE EKEFAEAKMK EYEASVSTKE VDPGKASKAA WIRKIKGLPI DNFMKEGKTA APELGQNVFI
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.