missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Ephrin-A2 Fc Chimera Biotinylated Protein

Product Code. 18327767
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18327767 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18327767 Supplier R&D Systems™ Supplier No. BT603

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Ephrin-A2 Fc Chimera Biotinylated Protein is derived from NS0. The Recombinant Mouse Ephrin-A2 Fc Chimera Biotinylated Protein has been validated for the following applications: Binding Activity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Gene ID (Entrez) 13637
Molecular Weight (g/mol) 46 kDa (monomer)
Name Ephrin-A2 Fc Chimera
Quantity 25 μg
Source Mouse myeloma cell line,NS0-derived mouse Ephrin-A2 protein, (N-terminus) Human Ephrin-A2 (Glu23-Asn184) Accession &Num; P52801 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Biotin
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL with sterile PBS.
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.