missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse EphA2 Fc Chimera Protein

Product Code. 18345557
Change view
Click to view available options
Quantity:
200 μg
Unit Size:
200µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18345557 200 μg 200µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18345557 Supplier R&D Systems™ Supplier No. 639A2200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Measured by its ability to bind Recombinant Mouse Ephrin-A1 Fc Chimera (Catalog # 602-A1 ) in a functional ELISA. Immobilized Recombinant Mouse EphA2 Fc Chimera at 2 μg/mL (100 μL/well) can bind Recombinant Mouse Ephrin-A1 Fc Chimera with a linear range of 0.078-5 ng/mL. Optimal dilutions should be determined by each laboratory for each application.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AAA82113
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris-HCl and EDTA.
Gene ID (Entrez) 1969
Molecular Weight (g/mol) M.W. (Observed): 105 kDa, reducing conditions; M.W. (theoretical): 84 kDa (monomer)
Name EphA2 Fc Chimera
Quantity 200 μg
Source Mouse myeloma cell line, NS0-derived mouse EphA2 protein Mouse EphA2 (Ala22-Ala535) (N-terminus) Accession # AAA82113 IEGRMD Human IgG1 (Pro100-Lys330) 6-His tag (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Endotoxin Concentration <1.0 EU / 1 μg of the protein by the LAL method.
Gene Symbol Epha2
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 100 μg/mL in sterile PBS.
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein EphA2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.