missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse Endoglycan/PODXL2 Protein

Product Code. 18796677
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18796677 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18796677 Supplier R&D Systems™ Supplier No. 8365EG050
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse Endoglycan/PODXL2 Protein is derived from NS0. The Recombinant Mouse Endoglycan/PODXL2 Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number XP_006506309
Formulation Lyophilized from a 0.2μm filtered solution in PBS
Gene ID (Entrez) 319655
Molecular Weight (g/mol) 51kDa
Name Endoglycan/PODXL2
Preparation Method Reconstitute at 100μg/mL in PBS.
Quality Control Testing SDS-PAGE: 90-124kDa, reducing conditions
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse Endoglycan/PODXL2 protein Gly30-Gly498,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Regulatory Status RUO
Termination Gly30
Without Additives Carrier Protein
Conjugate Unconjugated
Recombinant Recombinant
Product Line R&D Systems
Purity or Quality Grade 95%, by SDS-PAGE with silver staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.