missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse EGF / Epidermal Growth Factor Protein

Product Code. 15925579
Change view
Click to view available options
Quantity:
1 mg
100 μg
5 mg
500 μg
Unit Size:
100µg
1mg
500µg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15925579 1 mg 1mg
15915579 100 μg 100µg
15935579 500 μg 500µg
15945579 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15925579 Supplier enQuireBio™ Supplier No. QP40081mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Specifications

Accession Number NP_034243.2
Gene ID (Entrez) 13645
Name EGF / Epidermal Growth Factor Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Gene Symbol EGF
Biological Activity Shows bioactivity at a concentration of 250 pg/mL in the BALB/c 3T3 cell proliferation assay. The bioactivity acceptance criteria is set at 4 x 10^6 units/mg.
Product Type Recombinant Protein
Cross Reactivity Mouse
Species E. coli
Protein Tag Untagged
Sequence NSYPGCPSSY DGYCLNGGVC MHIESLDSYT CNCVIGYSGD RCQTRDLRWW ELR
Buffer 10 mM sodium phosphate, pH 7.5
Purity or Quality Grade Greater than 95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.