missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse EGF / Epidermal Growth Factor Protein
Specifications
Specifications
| Accession Number | NP_034243.2 |
| Gene ID (Entrez) | 13645 |
| Name | EGF / Epidermal Growth Factor Protein |
| Quantity | 100 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
| Gene Symbol | EGF |
| Biological Activity | Shows bioactivity at a concentration of 250 pg/mL in the BALB/c 3T3 cell proliferation assay. The bioactivity acceptance criteria is set at 4 x 10^6 units/mg. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?