missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse EG VEGF Protein
A cDNA sequence encoding the EG VEGF was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11757-20ug
Additional Details : Weight : 0.01000kg
Specifications
EG VEGF Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
AVITGACERD IQCGAGTCCA ISLWLRGLRL CTPLGREGEE CHPGSHKIPF LRKRQHHTCP CSPSLLCSRF PDGRYRCFRD LKNANF | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Mouse | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS pH 7.4 and 3% Trehalose. |