missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse EDA-A1/Ectodysplasin A1 Protein

Product Code. 18328777
Change view
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
10µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Formulation unitSize
18328777 Lyophilized from a 0.2μm filtered solution in PBS. 10µg
18178140 Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18328777 Supplier R&D Systems™ Supplier No. 191ED010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

The Recombinant Mouse EDA-A1/Ectodysplasin A1 Protein is derived from NS0. The Recombinant Mouse EDA-A1/Ectodysplasin A1 Protein has been validated for the following applications: Binding Activity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 13607
Molecular Weight (g/mol) 23 kDa
Name EDA-A1/Ectodysplasin A1
Quantity 10 μg
Source Mouse myeloma cell line,NS0-derived mouse EDA-A1/Ectodysplasin A1 protein Ala179-Ser391,with an N-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Homotrimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.