missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse E-Cadherin Protein

Product Code. 18798707
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18798707

Brand: R&D Systems™ 8875EC050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Mouse E-Cadherin Protein is derived from NS0. The Recombinant Mouse E-Cadherin Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P09803
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in MES, NaCl and CaCl 2 .
Gene ID (Entrez) 999
Molecular Weight (g/mol) Observed M.W.: 75-87kDa, reducing conditions, Theoretical M.W.: 62kDa
Quantity 50 μg
Source Mouse myeloma cell line,NS0-derived mouse E-Cadherin protein Asp157-Val709,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias Arc-1, CAD1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDH1, CDHE, cell-CAM 120/80, Cell-CAM120/80, ECAD, E-cadherin, ECadherin, Epithelial cadherin, LCAM, L-CAM, UVOE-Cadherin, uvomorulin
Common Name E-Cadherin
Gene Symbol Cdh1
Biological Activity Measured by the ability of the immobilized protein to support the adhesion of the MCF-7 human breast cancer cells. The ED50 for this effect is typically 0.25-1μg/mL.
Conjugate Unconjugated
Species Mouse
Recombinant Recombinant
Expression System Mouse myeloma cell line, NS0-derived Asp157-Val709, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE with Silver Staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.