missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse DNAM-1/CD226 Fc Chimera Protein

Product Code. 18375627
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18375627 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18375627 Supplier R&D Systems™ Supplier No. 4436DN050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Binding Activity

Measured by its binding ability in a functional ELISA. When Recombinant Mouse Nectin-2/CD112 (Catalog # 3869-N2) is coated at 1 μg/mL, Recombinant Mouse DNAM-1/CD226 Fc Chimera binds with a typical ED50 of 0.3-1.8 μg/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8K4F0
For Use With (Application) Binding Activity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 10666
Molecular Weight (g/mol) M.W. (Observed): 70-90 kDa, reducing conditions; M.W. (theoretical): 53.9 kDa
Name DNAM-1/CD226 Fc Chimera
Quantity 50 μg
Source Mouse myeloma cell line, NS0-derived mouse DNAM-1/CD226 protein Mouse DNAM-1/CD226 (Glu19-Pro254) (N-terminus) Accession # Q8K4F0 IEGRMDP Mouse IgG2A (Glu98-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.01 EU / 1 μg of the protein by the LAL method.
Gene Symbol Cd226
Conjugate Unconjugated
Recombinant Recombinant
Reconstitution Reconstitute at 500 μg/mL in sterile PBS.
Purity or Quality Grade >80%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein DNAM-1/CD226
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.