missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Desert Hedgehog C23II protein is fully biologically active when compared to standard. The ED50 as determined by its ability to induce alkaline phosphatase production by murine MC3T3E1 cells is 5-20 μg/mL.
Specifications
Specifications
| Accession Number | Q61488, Q61488, Q61488 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4, 1 mM DTT, 0.05 % Tween-80. |
| Gene ID (Entrez) | 50846 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 20.0 kDa |
| Quantity | 5 μg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/μg of Desert Hedgehog C23II as determined by LAL method. |
| Gene Symbol | DHH |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?