missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Recombinant Mouse Desert Hedgehog C23II Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
1mg
5 μg
Unit Size:
100µg
1mg
5µg
Description
Desert Hedgehog C23II protein is fully biologically active when compared to standard. The ED50 as determined by its ability to induce alkaline phosphatase production by murine MC3T3E1 cells is 5-20 μg/mL.
Specifications
Specifications
| Accession Number | Q61488, Q61488, Q61488 |
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4, 1 mM DTT, 0.05 % Tween-80. |
| Gene ID (Entrez) | 50846 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 20.0 kDa |
| Quantity | 1mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 0.1 EU/μg of Desert Hedgehog C23II as determined by LAL method. |
| Gene Symbol | DHH |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction