missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Defensin beta 2 protein is fully biologically active when compared to standard. The biologically active determined by a chemotaxis bioassay using immature human dendritic cells is in a concentration of 10-100 ng/mL.
Specifications
Specifications
| For Use With (Application) | PAGE, Bioactivity |
| Formulation | Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4. |
| Gene ID (Entrez) | 13215 |
| Molecular Weight (g/mol) | M.W. (Theoretical): 5.5 kDa |
| Quantity | 1mg |
| Storage Requirements | Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles. |
| Endotoxin Concentration | Less than 1 EU/μg of Defensin beta 2 as determined by LAL method. |
| Gene Alias | BD-2, Defb2, Defb2 defensin beta 2, defensin beta 2 |
| Reconstitution | Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C |
| Research Category | Biologically Active Proteins |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?