missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse D-dopachrome decarboxylase Protein
A DNA sequence encoding the Mus musculus (Mouse) D-dopachrome decarboxylase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5921-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_034157.1 | |
D-dopachrome decarboxylase Protein | |
Research Use Only | |
Ddt | |
Recombinant Protein | |
E. coli | |
PFVELETNLP ASRIPAGLEN RLCAATATIL DKPEDRVSVT IRPGMTLLMN KSTEPCAHLL VSSIGVVGTA EQNRTHSASF FKFLTEELSL DQDRIVIRFF PLEAWQIGKK GTVMTFL | |
Greater than 90% as determined by SDS-PAGE. |
13202 | |
500 μg | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |