missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Mouse CXCL9/MIG Protein

Product Code. 18477391 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
1mg
5 μg
500 μg
Unit Size:
100µg
1mg
500µg
5µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18477391 100 μg 100µg
18490222 1mg 1mg
18416501 500 μg 500µg
18420352 5 μg 5µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18477391 Supplier Novus Biologicals™ Supplier No. NBP235191100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

MIG Protein is fully biologically active when compared to standard. The biological activity determined by a chemotaxis bioassay using human lymphocytes is in a concentration range of 0.1-1.0 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P18340, P18340, P18340
For Use With (Application) PAGE, Bioactivity
Formulation Lyophilized from a 0.2 um filtered concentrated solution in 2 x PBS, pH 7.4.
Gene ID (Entrez) 4283
Molecular Weight (g/mol) M.W. (Theoretical): 12.2 kDa
Quantity 100 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/μg of CXCL9/MIG as determined by LAL method.
Gene Alias chemokine (C-X-C motif) ligand 9, CMK, crg-10, C-X-C motif chemokine 9, CXCl9, Gamma-interferon-induced monokine, Humig, MIG/CXCL9, MIGSmall-inducible cytokine B9, monokine induced by gamma interferon, SCYB9Monokine induced by interferon-gamma
Gene Symbol CXCL9
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C
Research Category Biologically Active Proteins, Chemokines and Cytokines, Signal Transduction
Purity or Quality Grade >95%, by SDS-PAGE and HPLC
Protein CXCL9/MIG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.