missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CXCL7/Thymus Chemokine-1 (aa 48-109)

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

Brand:  R&D Systems™ 1091-CK-025/CF

Additional Details : Weight : 0.00970kg

 View more versions of this product

Product Code. 18127340

  • £327.21 / 25µg
Estimated Shipment: 04-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Recombinant Mouse CXCL7/Thymus Chemokine-1 (aa 48-109) is derived from E. coli. The Recombinant Mouse CXCL7/Thymus Chemokine-1 (aa 48-109) has been validated for the following applications: Bioactivity.
Specifications

Specifications

Lyophilized from a 0.2μm filtered solution in PBS.
6.8 kDa
25μg
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Recombinant
57349
CXCL7/Thymus Chemokine-1 aa 48-109
E. coli-derived mouse CXCL7/Thymus Chemokine-1 protein Ile48-Ile109
Unconjugated
97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers