missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse CXCL3/DCIP-1 Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
£343.40
Spécification
Gene ID (Entrez) | 330122 |
---|---|
Molecular Weight (g/mol) | 7.9 kDa |
Name | CXCL3/GRO gamma/CINC-2/DCIP-1 |
Quantity | 25μg |
Source | E. coli-derived mouse CXCL3/GRO gamma/CINC-2/DCIP-1 protein Ala28-Ser100 |
Code produit | Marque | Formulation | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Formulation | Prix | Quantité et disponibilité | |||||
18176250
|
R&D Systems™
5568-CA-025 |
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. |
£343.40
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18187830
|
R&D Systems™
5568-CA-025/CF |
Lyophilized from a 0.2μm filtered solution in PBS. |
£343.40
25µg |
Estimated Shipment: 21-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The Recombinant Mouse CXCL3/DCIP-1 Protein is derived from E. coli. The Recombinant Mouse CXCL3/DCIP-1 Protein has been validated for the following applications: Bioactivity.Spécification
330122 | |
CXCL3/GRO gamma/CINC-2/DCIP-1 | |
E. coli-derived mouse CXCL3/GRO gamma/CINC-2/DCIP-1 protein Ala28-Ser100 | |
Unconjugated | |
95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
7.9 kDa | |
25μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |