missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse CXCL16 (Histidine-tagged) Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.
£211.66
Specifications
Accession Number | Q8BSU2.2 |
---|---|
For Use With (Application) | Bioactivity |
Gene ID (Entrez) | 58191 |
Molecular Weight (g/mol) | M.W. (Observed): 35-40 kDa, reducing conditions; M.W. (theoretical): 19.9 kDa |
Name | CXCL16 Extracellular Domain |
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Formulation | Price | Quantity & Availability | |||||
18146200
|
R&D Systems™
1165-CX-025/CF |
Lyophilized from a 0.2 μm filtered solution in PBS. |
£211.66
25µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18166980
|
R&D Systems™
1165-CX-025 |
Lyophilized from a 0.2 μm filtered solution in PBS with BSA as a carrier protein. |
£211.66
25µg |
Estimated Shipment: 04-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Measured by its ability to chemoattract BaF3 mouse pro-B cells transfected with mouse CXCR6. Matloubian, M. et al. (2000) Nat. Immunol. 1:298. The ED50 for this effect is 4-20 ng/mL.Specifications
Q8BSU2.2 | |
58191 | |
CXCL16 Extracellular Domain | |
Mouse myeloma cell line, NS0-derived mouse CXCL16 protein Asn27-Trp201, with a C-terminal 6-His tag | |
<0.10 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining. |
Bioactivity | |
M.W. (Observed): 35-40 kDa, reducing conditions; M.W. (theoretical): 19.9 kDa | |
25 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after reconstitution. | |
Cxcl16 | |
Recombinant | |
CXCL16 |