missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse CXCL12 / SDF-1 Isoform beta Protein
Recombinant Protein
£80.80 - £3085.00
Specifications
Accession Number | NP_038683.1 |
---|---|
Name | CXCL12 / SDF-1 Isoform beta Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 1.00 EUs/ug as determined by the LAL method. |
Gene Symbol | CXCL12 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15985709
|
enQuireBio™
QP5216-2UG |
2 μg |
£80.80
2µg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15965709
|
enQuireBio™
QP5216-10UG |
10 μg |
£191.00
10µg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955709
|
enQuireBio™
QP5216-100UG |
100 μg |
£476.00
100µg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15995709
|
enQuireBio™
QP5216-500UG |
500 μg |
£1890.00
500µg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15975709
|
enQuireBio™
QP5216-1MG |
1 mg |
£3085.00
1mg |
Estimated Shipment: 29-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_038683.1 | |
Research Use Only | |
CXCL12 | |
Recombinant Protein | |
E. coli | |
KPVSLSYRCP CRFFESHIAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRL KM | |
Greater than 95% as determined by SDS-PAGE. |
CXCL12 / SDF-1 Isoform beta Protein | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Mouse CXCL12 / SDF-1 Isoform beta induces primary human T cell chemotaxis (significanty different from control at Mouse CXCL12 / SDF-1 Isoform beta concentrations greater than 5-50 ng/ml). | |
Mouse | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |