missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Mouse CXCL1/KC Protein

Product Code. 18187820
Change view
Click to view available options
Quantity:
10 μg
50 μg
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
10µg
50µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Formulation unitSize
18187820 10 μg Lyophilized from a 0.2μm filtered solution in PBS. 10µg
18375617 10 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 10µg
18116310 50 μg Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein. 50µg
18318387 50 μg Lyophilized from a 0.2μm filtered solution in PBS. 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18187820 Supplier R&D Systems™ Supplier No. 453KC010/CF

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Mouse CXCL1/KC Protein is derived from E. coli.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 14825
Molecular Weight (g/mol) 8 kDa
Name CXCL1/GRO alpha/KC/CINC-1 aa 20-96
Quantity 10 μg
Source E. coli-derived mouse CXCL1/GRO alpha/KC/CINC-1 protein Arg20-Lys96
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Gene Alias CINC1, CINC-1, FSP, GRO alpha, GRO1, GROa, KC, MGSA, MGSA-a, MGSA-alpha, NAP-3, SCYB1
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 97%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.