missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Mouse CX3CL1/Fractalkine (Full Length) Protein
Click to view available options
Formulation:
Lyophilized from a 0.2μm filtered solution in PBS with BSA as a carrier protein.
Lyophilized from a 0.2μm filtered solution in PBS.
Unit Size:
25µg
Description
The Recombinant Mouse CX3CL1/Fractalkine (Full Length) is derived from Sf 21 (baculovirus). The Recombinant Mouse CX3CL1/Fractalkine (Full Length) has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in PBS. |
| Gene ID (Entrez) | 20312 |
| Molecular Weight (g/mol) | 34 kDa |
| Name | CX3CL1/Fractalkine Full Length |
| Quantity | 25 μg |
| Source | Spodoptera frugiperda,Sf 21 (baculovirus)-derived mouse CX3CL1/Fractalkine protein Gln25-Arg337,with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.1 EU per 1μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction